Skip to Content
Merck

Skip To

BDPH9BCC9C75

Bide Pharmatech

3-Chloro-4-(3-(trifluoromethyl)phenoxy)aniline

Synonym(s):

3-chloro-4-[3-(trifluoromethyl)phenoxy]aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C13H9O1N1Cl1F3
CAS Number:
Molecular Weight:
287.67
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H9ClF3NO/c14-11-7-9(18)4-5-12(11)19-10-3-1-2-8(6-10)13(15,16)17/h1-7H,18H2

Inchi Key

WTPFYPUXUHYGIP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library