Skip to Content

BDPH9BCCBC6B

Bide Pharmatech

4-Chloro-1,10-phenanthroline

Synonym(s):

4-chloro[1,10]phenanthroline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H7N2Cl1
CAS Number:
Molecular Weight:
214.65
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H7ClN2/c13-10-5-7-15-12-9(10)4-3-8-2-1-6-14-11(8)12/h1-7H

Inchi Key

BAEVILLEIGDCDN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-Chloro-1,10-phenanthroline




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library