Skip to Content

BDPH9BCCF270

Bide Pharmatech

3-Iodo-1-methylpyridin-2(1H)-one

Synonym(s):

3-iodo-1-methylpyridin-2(1H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O1N1I1
CAS Number:
Molecular Weight:
235.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6INO/c1-8-4-2-3-5(7)6(8)9/h2-4H,1H3

Inchi Key

RMGDJLOUEQWPOU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-Solid or Liquid or Lump



Still not finding the right product?

Explore all of our products under 3-Iodo-1-methylpyridin-2(1H)-one




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library