Skip to Content

BDPH9BCD3DD5

Bide Pharmatech

7-Bromo-2-chloro-3-methylquinoline

Synonym(s):

7-bromo-2-chloro-3-methylquinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7N1Cl1Br1
CAS Number:
Molecular Weight:
256.53
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H7BrClN/c1-6-4-7-2-3-8(11)5-9(7)13-10(6)12/h2-5H,1H3

Inchi Key

SQFVOHFUJOPVEH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 7-Bromo-2-chloro-3-methylquinoline


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library