Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C8H8Cl2O2S/c1-6-4-7(2-3-8(6)9)5-13(10,11)12/h2-4H,5H2,1H3
Inchi Key
OIWQZLQYWYCVCP-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (4-Chloro-3-methylphenyl)methanesulfonyl chloride
hstatements
signalword
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

