Skip to Content
Merck

BDPH9BCD45F3

Bide Pharmatech

6-Chloro-5-methylpyridazin-3-amine

Synonym(s):

6-chloro-5-methyl-3-pyridazinamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H6N3Cl1
CAS Number:
Molecular Weight:
143.58
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H6ClN3/c1-3-2-4(7)8-9-5(3)6/h2H,1H3,(H2,7,8)

Inchi Key

UWDLNRUFHRYMSE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 6-Chloro-5-methylpyridazin-3-amine


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library