Skip to Content

BDPH9BCD4B69

Bide Pharmatech

3-Bromo-2-(trifluoromethyl)benzaldehyde

Synonym(s):

3-bromo-2-(trifluoromethyl)benzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H4O1Br1F3
CAS Number:
Molecular Weight:
253.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H4BrF3O/c9-6-3-1-2-5(4-13)7(6)8(10,11)12/h1-4H

Inchi Key

GNJZENBMBWRNAY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid or lump



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library