Skip to Content

BDPH9BCD6392

Bide Pharmatech

5-(Methylsulfonyl)pyridin-2(1H)-one

Synonym(s):

5-(methylsulfonyl)-2-pyridinol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H7O3N1S1
CAS Number:
Molecular Weight:
173.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H7NO3S/c1-11(9,10)5-2-3-6(8)7-4-5/h2-4H,1H3,(H,7,8)

Inchi Key

XFQLRNGQOCXUNF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library