Skip to Content

BDPH9BCDBACC

Bide Pharmatech

6-Bromo-2-chloro-3-fluoropyridine

Synonym(s):

6-bromo-2-chloro-3-fluoropyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H2N1Cl1Br1F1
CAS Number:
Molecular Weight:
210.43
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H2BrClFN/c6-4-2-1-3(8)5(7)9-4/h1-2H

Inchi Key

KPQMWTHSSFZPMJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Inert atmosphere, 2-8C



Still not finding the right product?

Explore all of our products under 6-Bromo-2-chloro-3-fluoropyridine


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library