Skip to Content
Merck

BDPH9BCDCED0

Bide Pharmatech

1-Benzyl-1H-pyrazole-5-carboxylic acid

Synonym(s):

1-benzyl-1H-pyrazole-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H10O2N2
CAS Number:
Molecular Weight:
202.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H10N2O2/c14-11(15)10-6-7-12-13(10)8-9-4-2-1-3-5-9/h1-7H,8H2,(H,14,15)

Inchi Key

SFIGVCZYZFTEMK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library