Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C4H2BrClIN3/c5-1-4(8)10-2(6)3(7)9-1/h(H2,8,10)
Inchi Key
YXQYTWWGBBHZBS-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Yellow solid
Still not finding the right product?
Explore all of our products under 3-Bromo-6-chloro-5-iodopyrazin-2-amine
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
