Skip to Content
Merck

BDPH9BCE1898

Bide Pharmatech

3-(3-Bromo-1H-pyrazol-1-yl)pyridine

Synonym(s):

3-(3-bromo-1H-pyrazol-1-yl)pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H6N3Br1
CAS Number:
Molecular Weight:
224.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6BrN3/c9-8-3-5-12(11-8)7-2-1-4-10-6-7/h1-6H

Inchi Key

AHFSEQPINGKCIQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library