Skip to Content
Merck

BDPH9C01F93B

Bide Pharmatech

trans-1-Bromo-3-(trifluoromethyl)cyclobutane

Synonym(s):

(1r,3r)-1-bromo-3-(trifluoromethyl)cyclobutane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H6Br1F3
CAS Number:
Molecular Weight:
203
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H6BrF3/c6-4-1-3(2-4)5(7,8)9/h3-4H,1-2H2/t3-,4-

Inchi Key

LHJMYJBLOMVORH-JPYJGEKTSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library