Skip to Content
Merck

BDPH9C0239AD

Bide Pharmatech

2-(Difluoromethoxy)naphthalene

Synonym(s):

2-(difluoromethoxy)naphthalene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H8O1F2
CAS Number:
Molecular Weight:
194.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H8F2O/c12-11(13)14-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H

Inchi Key

PHRDBYLNQKVKBB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to white to off-white solid or semi-solid or liquid



Still not finding the right product?

Explore all of our products under 2-(Difluoromethoxy)naphthalene


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library