Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Ships in 10 days - from Aldrich Partner.
| 288,00 PLN |
250 mg | Ships in 10 days - from Aldrich Partner.
| 360,00 PLN |
1 g | Ships in 10 days - from Aldrich Partner.
| 810,00 PLN |
About This Item
Vendor SKU: BD238673-100MG
288,00 PLN
Ships in 10 days - from Aldrich Partner.
Inchi Code
1S/C8H8BrNO3/c1-5-6(9)3-4-7(13-2)8(5)10(11)12/h3-4H,1-2H3
Inchi Key
RZOPBDDNOSELGK-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 1-Bromo-4-methoxy-2-methyl-3-nitrobenzene
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
