Skip to Content
Merck

Skip To

C-141S

Cannabigerol (CBG) solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Cannabigerol solution, CBG solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
1.600,00 PLN

About This Item

Empirical Formula (Hill Notation):
C21H32O2
CAS Number:
Molecular Weight:
316.48
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21
eCl@ss:
25140904

1.600,00 PLN


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material

feature

second source lots, Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

application(s)

forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CCCCCC1=CC(O)=C(C/C=C(C)/CCC=C(C)C)C(O)=C1

InChI

1S/C21H32O2/c1-5-6-7-11-18-14-20(22)19(21(23)15-18)13-12-17(4)10-8-9-16(2)3/h9,12,14-15,22-23H,5-8,10-11,13H2,1-4H3/b17-12+

InChI key

QXACEHWTBCFNSA-SFQUDFHCSA-N

General description

A non-psychoactive cannabinoid found in Cannabis, Cannabigerol (CBG) reportedly has efficacy for medical conditions such as glaucoma and inflammatory bowel disease. This Certified Spiking Solution® is second source standard suitable for cannabigerol testing methods by HPLC, GC/MS or LC-MS/MS for forensic analysis, clinical toxicology, urine drug testing, and Cannabis potency or impurity profiling testing applications. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C141PHL85956C212
description

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

description

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

description

phyproof® Reference Substance

description

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

application(s)

forensics and toxicology

application(s)

cannabis testing

application(s)

cannabis testing

application(s)

cannabis testing

format

single component solution

format

single component solution

format

-

format

single component solution

grade

certified reference material

grade

certified reference material

grade

primary reference standard

grade

certified reference material

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

Quality Level

300

Quality Level

300

Quality Level

-

Quality Level

-

concentration

1 mg/mL in methanol

concentration

1.0 mg/mL in methanol

concentration

-

concentration

100 μg/mL in methanol


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library