Skip to Content
Merck

Skip To

NMIS004

5α-Androstan-3α,17β-diol-O-3-β-glucuronic acid

NMI Australia

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C25H40O8
CAS Number:
Molecular Weight:
468.58
UNSPSC Code:
41116107
eCl@ss:
25140904


agency

NMI Australia certified reference material

manufacturer/tradename

NMI Australia

format

neat

storage temp.

2-8°C

InChI

1S/C25H40O8/c1-24-9-7-13(32-23-20(29)18(27)19(28)21(33-23)22(30)31)11-12(24)3-4-14-15-5-6-17(26)25(15,2)10-8-16(14)24/h12-21,23,26-29H,3-11H2,1-2H3,(H,30,31)/t12?,13?,14?,15?,16?,17-,18-,19+,20-,21-,23-,24+,25+/m1/s1

InChI key

GYNWSIBKBBWJJW-ZQMNIGHUSA-N



Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters