Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C20H32O2
CAS Number:
Molecular Weight:
304.47
UNSPSC Code:
41116107
grade
certified reference material
agency
NMI Australia certified reference material
form
solid
manufacturer/tradename
NMI Australia
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
neat
storage temp.
2-8°C
InChI
1S/C20H32O2/c1-12-11-20(3)13(10-17(12)21)4-5-14-15-6-7-18(22)19(15,2)9-8-16(14)20/h12-16,18,22H,4-11H2,1-3H3/t12-,13+,14+,15+,16+,18+,19+,20+/m1/s1
InChI key
IKXILDNPCZPPRV-RFMGOVQKSA-N
signalword
Danger
hcodes
Hazard Classifications
Carc. 2 - Repr. 1B
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
