Skip to Content
Merck

COM448681237

Combi-Blocks, Inc.

3-(2-Oxo-1,3-thiazolidin-3-yl)propanoic acid

Synonym(s):

3-(2-oxo-1,3-thiazolidin-3-yl)propanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
175.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H9NO3S/c8-5(9)1-2-7-3-4-11-6(7)10/h1-4H2,(H,8,9)

Inchi Key

XJXGOBWSTJVANH-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN



Still not finding the right product?


MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?ST-5030



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library