Skip to Content
Merck

AI Assist - BETA


Expedite your answers with the help of AI

73103BC

GDS RGQ Laptop US Power Cord & Charger - USB Type C flat

BioControl Systems, for use with GDSlaptop

Synonym(s):

Laptop Charger Cord
GDS Laptop Power Cable US and Charger
GDS Laptop AC Adapter and Charger
GDS Laptop Power Supply US and Charger
GDS Laptop Charging Cable US and Charger
GDS Laptop Power Cord for US Outlet and Charger
GDS Laptop

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116107
NACRES:
NE.02
eCl@ss:
32035490
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 unit

AI Assist - BETA


General description

The GDS Laptop is an essential part of theAssurance® GDS Rotor-Gene Q thermocycler. The GDS Laptop Power Cordis suitable for use with Latitude 5430 Rugged, 90W charger laptop models in theNA region.Assurance®GDS is an automated nucleic acid amplification system for the detection ofpathogens in food, ingredients, and environmental samples.

Application

GDS Laptop Power Cord US and Charger areessential accessories to be used with the GDS Rotor-Gene Q laptop provided withthe Assurance® GDS system.

Legal Information

Assurance GDS is a registered trademark of BioControl Systems, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library