Skip to Content
Merck

ABC22

Anti-Atg4D Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cysteine protease ATG4D, AUT-like 4 cysteine endopeptidase, Autophagin-4, Autophagy-related cysteine endopeptidase 4, Autophagy-related protein 4 homolog D

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATG4D(84971)

General description

Atg4D (autophagy-related protein 4D) is a member of the Atg4 family of C54 endopeptidases that are required for the synthesis of autophagosomes. Atg4D is found to be cleaved by caspase-3 into a more active form where it can interact with the autophagosome marker Atg8. Atg4D is also known to be associated with mitochondrial dysfunction and the induction of apoptosis.
~56 kDa observed.
The calculated molecular weight is 53 kDa; however, Atg4D has been shown as a ~55 kDa band in western blots (Li, M., et al. (2011). J Biol Chem. 286(9):7327-7338.)

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human Atg4D.

Application

Anti-Atg4D Antibody is an antibody against Atg4D for use in WB, IH(P).
Immunohistochemistry (Paraffin) Analysis: A 1:200 dilution of a previous lot detected Atg4D in human testicular cancer tissue.

Western Blot Analysis: 1 µg/mL from a representative lot detected Atg4D on 10 µg of human fetal skeletal muscle tissue, NIH/3T3, and L6 cell lysates.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

This antibody recognizes Atg4D at the N-terminus.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate
Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 0.05 µg/mL of this antibody detected Atg4D in 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library