Skip to Content
Merck

Skip To

ABC829

Anti-SCRN1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Secernin-1, SCRN1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on13 November 2026fromKuehne + Nagel Sp. z o.o.

2.040,00 PLN

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC, WB
Citations:
-

2.040,00 PLN


Estimated to ship on13 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SCRN1(9805)

General description

Secernin-1 (SCRN1) is involved in the regulation of exocytosis from peritoneal mast cells. More recent studies have shown that SCRN1 expression is upregulated in gastric cancer cell lines and may possess epitopes that could function as tumor-associated antigens, potentially providing targets for cancer vaccines in the treatment of gastric cancers. Another report indicates that decreased expression of SCRN1 via RNAi expression resulted in significantly lower rates of cell growth in colorectal cancer cell lines, and increased SCRN1 expression in patients with colorectal cancer correlated with poor prognosis, suggesting that SCRN1 may also be involved in the regulation of cell growth and might be useful as a prognostic tool.
~30 and ~48 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: C-terminus
KLH-conjugated synthetic peptide corresponding to the C-terminus of human SCRN1.

Application

Anti-SCRN1 Antibody is an antibody against SCRN1 for use in western blotting, IHC & immunofluorescence.
Immunohistochemistry Analysis: 5 μg/mL from a representative lot detected SCRN1 in human kidney tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected SCRN1 in human kidney cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human kidney tissue with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected SCRN1 in human kidney tissue with and without blocking peptide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA022934HPA034690HPA023434
description

from rabbit, purified by affinity chromatography

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


WGK

WGK 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters