Skip to Content
Merck

Skip To

ABC930

Anti-PATE1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Prostate and testis expressed protein 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship on12 November 2026fromKuehne + Nagel Sp. z o.o.

1.850,00 PLN

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2

1.850,00 PLN


Estimated to ship on12 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PATE1(160065)

General description

Prostate and testis expressed protein 1 (PATE1) is a secreted protein that, as the name suggests, is expressed exclusively in the prostate and testis. PATE1 is up regulated in the prostate epithelial cells during cancer, and its extreme specificity of expression makes it a potential target for immunotherapy in prostate cancer. PATE1 is the founding member of new syntenic loci in humans and mice that encode secreted proteins that have 10 cysteine residues. The PATE family has three-fingered protein/Ly-6/urokinase-type plasminogen activator receptor (uPAR) domains that shape three-fingered proteins (TFPs). Additional members include PATE-M, PATE-DJ, and PATE-B that are also expressed in prostate and testis and sperm. Related proteins, PATE-P and PATE-Q are expressed only in placenta. Research indicates that PATE proteins may have something to do with fertility and recent research showed that addition PATE-like genes were expressed in neuron-rich tissues and thus may have a role in neurological function or transmission.
~14 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

GST-tagged recombinant protein corresponding to human PATE1.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected PATE1 in human testis and human prostate tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-PATE1 Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of PATE1.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected PATE1 in 10 µg of PC3 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABC244HPA076587ABC522
description

from rabbit, purified by affinity chromatography

description

from rabbit, purified by affinity chromatography

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

-

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

300

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-

Gene Information

human ... PATE1(160065)

Gene Information

human ... PATE1(160065)

Gene Information

-

Gene Information

-


Still not finding the right product?

Explore all of our products under Anti-PATE1 Antibody

— or —

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters