Skip to Content
Merck

Skip To

MABC554

Anti-Cullin-4A/CUL4A Antibody, clone 2A2.1

clone 2A2.1, from mouse

Synonym(s):

Cullin-4A, CUL-4A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on10 November 2026fromKuehne + Nagel Sp. z o.o.

2.330,00 PLN

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
2A2.1, monoclonal
Application:
WB
Citations:
-

2.330,00 PLN


Estimated to ship on10 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A2.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CUL4A(8451)

General description

Cullin-4A (UniProt Q13619; also known as CUL-4A) is encoded by the CUL4A gene (Gene ID 8451) in human. Polyubiquitination of proteins is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type (really interesting new gene), HECT-type (homologous to E6-AP C-terminus), and U-box type of E3 ligases. Cullin-4 belongs to the cullin family of hydrophobic proteins that confer substrate specificity to the multimeric Cullin-RING E3 ubiquitin ligase (CRL) complexes by acting as scaffold proteins. The human genome encodes 8 cullin proteins (CUL 1, 2, 3, 4A, 4B, 5, 7 and 9). The activity of CRLs is regulated by Cullin Neddylation via the reversible conjugation of a small ubiquitin-type protein known as Neural precursor cell expressed developmentally down-regulated protein 8 (Nedd8). CUL4A interacts with the substrate-binding adaptor DDB1 (DNA damage binding protein 1) via its arc-shaped helical N-terminal domain, and binds the small RING finger protein ROC1 (regulator of cullins 1) via its globular C-terminal domain. Despite the 83% sequence identity and seemingly functional redundancy between CUL4A and CUL4B, much more focus is placed on CUL4A in oncogenesis research.
~88 kDa observed

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to human Cullin-4A/CUL4A near the C-terminus.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Cullin-4A/CUL4A Antibody, clone 2A2.1 is validated for use in Western Blotting for the detection of Cullin-4A/CUL4A.

Biochem/physiol Actions

Clone 2A2.1 is expected to react with both spliced isoforms of human cullin-4A

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human melanoma SK-MEL-28 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Cullin-4A/CUL4A in 10 µg of human melanoma SK-MEL-28 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5701586SAB6012516PLA0136
description

clone 2A2.1, from mouse

description

-

description

clone 23GB790, recombinant rabbit monoclonal, expressed in HEK293 cells

description

Powered by Bethyl Laboratories, Inc.

clone

2A2.1, monoclonal

clone

monoclonal

clone

23GB790, recombinant monoclonal

clone

polyclonal

species reactivity

human

species reactivity

rat, mouse, human

species reactivity

human

species reactivity

human, mouse

Gene Information

human ... CUL4A(8451)

Gene Information

human ... CUL4A(8451)

Gene Information

human ... CUL4A(8451)

Gene Information

rabbit ... Cul4a(8451)

biological source

mouse

biological source

rabbit

biological source

rabbit (Monoclonal)

biological source

rabbit

antibody form

purified immunoglobulin

antibody form

-

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters