Skip to Content
Merck

Skip To

MABE2005

Anti-RNF4 Antibody, clone 11-3.1

Synonym(s):

E3 ubiquitin-protein ligase RNF4, Protein SNURF, RING finger protein 4, Small nuclear ring finger protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg
Please contact Customer Service for Availability
815,00 PLN
100 μg
Please contact Customer Service for Availability
2.050,00 PLN

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
11-3.1, monoclonal
Application:
ICC, WB
Citations:
-

815,00 PLN


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

11-3.1, monoclonal

mol wt

calculated mol wt 21.32 kDa, observed mol wt ~32 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG2bκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RNF4(6047)

General description

E3 ubiquitin-protein ligase RNF4 (UniProt: P78317; also known as RING finger protein 4, EC:2.3.2.27, Small nuclear ring finger protein, Protein SNURF) is encoded by the RNF4 (also known as SNURF) gene (Gene ID: 6047) in human. RNF4 gene encodes ubiquitin E3 ligase, an enzyme widely expressed in many tissues. RNF4 is mapped to human chromosome 4p16.3. The protein contains a RING-type zinc finger domain and around four small ubiquitin-related modifier (SUMO)-interaction motifs. The RING-type zinc finger domain is required for the ubiquitination of polysumoylated substrates and the SUMO interaction motifs (SIMs) mediates the binding to polysumoylated substrate. RNF4 is predominantly localized in the nucleus but can also be found in the cytoplasm. This dual localization allows it to function in multiple cellular compartments, dealing with SUMOylated substrates wherever they are located. The movement between these compartments can be regulated by specific signals or stress conditions. RNF4 is broadly expressed across various tissues, with notable levels in tissues that have high cellular turnover or are particularly responsive to stress, such as in immune cells and testis. It is involved in DNA repair, in transcription and chromatin regulation. It is also associated with the process of hematopoiesis. RNF4 is implicated in acute promyelocytic leukemia, acute myeloid leukemia, Fanconi anemia and aplastic anemia. Its role in maintaining protein homeostasis and responding to cellular stress makes it a key player in health and disease, with its dysfunction being linked to cancer, neurodegeneration, and developmental issues. Understanding the mechanisms regulating RNF4 activity and localization can provide insights into therapeutic strategies for these diseases. (Ref.: Saito, M., et al. (2014). Biochem Biophys Res Commun. 447(1):83-8. (PMID: 24695317).

Immunogen

GST-tagged, recombinant human RNF4 protein.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution from a representative lot detected RNF4 protein in lysate from HeLa cells.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected RNF4 in Immunocytochemistry application (Saito, M., et al. (2014). Biochem Biophys Res Commun. 447(1):83-8).

Western Blotting Analysis: A representative lot detected RNF4 in Western Blotting application (Saito, M., et al. (2014). Biochem Biophys Res Commun. 447(1):83-8).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 11-3.1 is a mouse monoclonal antibody that specifically detects recombinant human RNF4 protein.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA049356HPA022047MABC606
description

-

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

clone 11C3.1, from mouse

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

clone

11-3.1, monoclonal

clone

polyclonal

clone

polyclonal

clone

11C3.1, monoclonal

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library