Skip to Content
Merck

Skip To

MABE232

Anti-IGF2BP2 Antibody, clone 5E10-3E1

clone 5E10-3E1, from mouse

Synonym(s):

Insulin-like growth factor 2 mRNA-binding protein 2, IGF2 mRNA-binding protein 2, IMP-2, Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, VICKZ family member 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
5E10-3E1, monoclonal
Application:
WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E10-3E1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IGF2BP2(10644)

General description

Insulin-like growth factor 2 mRNA-binding protein 2 (IGF2BP2) is a member of the IGF-II mRNA binding protein family that posttranscriptionally regulates IGF2 during developmental stages. IGF2 mRNA-binding protein family members (IGF2BP1, 2 and 3) bind UTR region of IGF2 transcripts and are implicated in the transport of RNA targets to enable protein synthesis at specific locations in the cytoplasm. Studies have indicated variations in the IGF2BP2 gene may affect fetal glucose metabolism and plays a role in type 2 diabetes. In addition to playing a role in glucose metabolism and dieabetes, the insulin-like growth factor (IGF) signaling axis plays an important role in cancer biology. Research has shown IGF2 to be emerging as an important gene for ovarian cancer.
~66 kDa observed. An uncharacterized band at ~50 kDa may be observed in some lysates. Alternative splicing can result in 5 isoforms with molecular weights ranging from 55-66 kDa

Immunogen

Histidine-tagged recombinant protein corresponding to human IGF2BP2.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-IGF2BP2 Antibody, clone 5E10-3E1 (mouse monoclonal antibody) validated in WB to detect IGF2BP2 also known as Insulin-like growth factor 2 mRNA-binding protein 2, IGF2 mRNA-binding protein 2, IMP-2, Hepatocellular carcinoma autoantigen p62.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate
Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: A 1:20,000 dilution of this antibody detected IGF2BP2 on 10 µg of HepG2 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5701598HPA021367ABC504
description

clone 5E10-3E1, from mouse

description

-

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

from rabbit, purified by affinity chromatography

Gene Information

human ... IGF2BP2(10644)

Gene Information

human ... IGF2BP2(10644)

Gene Information

human ... IGF2BP1(10642)

Gene Information

human ... IGF2(3481)

species reactivity

human

species reactivity

human, rat

species reactivity

human

species reactivity

human

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

-

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

clone

5E10-3E1, monoclonal

clone

monoclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters