Skip to Content
Merck

PG9H9BB3873E

PrecisePEG

Azido-PEG3-Thiol

Synonym(s):

2-(2-(2-(2-azidoethoxy)ethoxy)ethoxy)ethane-1-thiol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product PG9H9BB3873E is not currently sold in your country.


Inchi Code

1S/C8H17N3O3S/c9-11-10-1-2-12-3-4-13-5-6-14-7-8-15/h15H,1-8H2

Inchi Key

GAXDQYGDMXOVCJ-UHFFFAOYSA-N

Storage temp.

-20

Purity

96%

Country of Origin

CN

Shipping Temperature

ambient

Physical Form

Yellow



MSDS

https://precisepeg.com/cdn/shop/files/AG-1782_SDS.pdf

hstatements

pcodes



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library