Skip to Content
Merck

57740

Iodobenzene

puriss., ≥99.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H5I
CAS Number:
Molecular Weight:
204.01
EC Number:
209-719-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1446140
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

puriss.

assay

≥99.0% (GC)

contains

silver wool as stabilizer

refractive index

n20/D 1.62 (lit.), n20/D 1.620

bp

188 °C (lit.)

mp

−29 °C (lit.)

density

1.830 g/mL at 20 °C, 1.823 g/mL at 25 °C (lit.)

SMILES string

Ic1ccccc1

InChI

1S/C6H5I/c7-6-4-2-1-3-5-6/h1-5H

InChI key

SNHMUERNLJLMHN-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Iodobenzene




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library