Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
agency
(National Research Council of Canada (NRC))
manufacturer/tradename
National Research Council of Canada (NRC)
application(s)
food and beverages
format
matrix material
General description
Features and Benefits
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description certified reference material | description certified reference material | description certified reference material | description certified reference material |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| format matrix material | format matrix material | format (Calibration solution) | format matrix material |
| agency (National Research Council of Canada (NRC)) | agency (National Research Council of Canada (NRC)) | agency (National Research Council of Canada (NRC)) | agency traceable to (National Research Council of Canada (NRC)) |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) |
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
