Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Please contact Customer Service for Availability | 332,00 PLN |
5 g | Please contact Customer Service for Availability | 828,00 PLN |
1 kg | ||
10 kg |
About This Item
Quality Segment
product line
ISOM8™
assay
(HPLC), ≥98%
form
solid
mol wt
536.77
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Cross Couplings, reaction type: Fluorinations, reagent type: ligand
greener alternative product score
old score: 8
new score: 1
Find out more about DOZN™ Scoring
sustainability
Sustainability features
mp
191-194 °C (Lit.), 187-195 °C
application(s)
diagnostic assay manufacturing
life science and biopharma
functional group
phosphine
storage temp.
room temp
SMILES string
COc1c(P(C2CCCCC2)C3CCCCC3)c(c4c(C(C)C)cc(C(C)C)cc4C(C)C)c(OC)cc1
InChI
1S/C35H53O2P/c1-23(2)26-21-29(24(3)4)33(30(22-26)25(5)6)34-31(36-7)19-20-32(37-8)35(34)38(27-15-11-9-12-16-27)28-17-13-10-14-18-28/h19-25,27-28H,9-18H2,1-8H3
InChI key
WDVGNXKCFBOKDF-UHFFFAOYSA-N
General description
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard
Application
BrettPhos, developed by the Buchwald group, is a highly effective dialkylbiaryl phosphine ligand that enhances transition metal-catalyzed reactions with unique steric and electronic properties.
Notable characteristics & key uses:
- Steric & Electronic Properties: Its bulky biaryl structure ensures stability, selectivity, and reduced side reactions, while the electron-deficient phosphine group boosts catalytic reactivity.
- Metal Binding: Excellent for palladium and nickel-catalyzed processes like Suzuki and Buchwald-Hartwig reactions, stabilizing the metal center for improved efficiency.
- Catalyst Stability: Enhances longevity and prevents degradation, delivering higher yields and cost-effectiveness under challenging conditions.
- Versatility: Works across various solvents, temperatures, and reaction conditions, making it ideal for diverse synthetic processes.
- Cross-Coupling Reactions: Promotes efficient carbon-carbon bond formation with minimal by-products in Suzuki, Heck, and Sonogashira reactions.
- Diverse Substrate Compatibility: Performs well with challenging functional groups, enabling versatile synthetic pathways.
- Enhanced Reaction Conditions: Allows for milder, eco-friendly reaction conditions.
- Scalability: Suitable for both small-scale labs and large-scale industrial applications, ensuring consistent results.
- Recyclable Pd(acac)₂/BrettPhos/PEG-1000 System for Suzuki-Miyaura Coupling: This study presents a sustainable catalytic system for nitroarenes, emphasizing its efficiency, recyclability, and greener approach to C–C bond formation. (Mingzhong Cai et al., 2024).
- Pd-Catalyzed C–N Coupling via Buchwald–Hartwig amination using BrettPhos ligand: This study demonstrates the scalabilityand efficiency of BrettPhos in palladium-catalyzed C–N bond formation, enablingselective coupling under mild conditions suitable for the synthesis of complexmolecules on a larger scale. (Seb Caille et al., 2015).
- Pd-Catalyzed Denitrative Cyanation of Nitroarenes with BrettPhos Ligand: This study highlights the denitrative cyanation of nitroarenes using amino acetonitriles. The method efficiently converts nitroarenes into aryl nitriles, offering a halogen- and metal-waste-free catalytic process suitable for large-scale applications (Junichiro Yamaguchi et al., 2024).
Incorporating BrettPhos into your synthetic strategies enhances efficiency, reliability, and innovation, offering robust support for research applications while ensurin gseamless scalability for manufacturing.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98%, ISOM8™ | description 98% | description - | description AldrichCPR |
| reaction suitability reaction type: Cross Couplings, reagent type: ligand, reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Fluorinations | reaction suitability reagent type: ligand | reaction suitability reagent type: catalyst | reaction suitability - |
| assay (HPLC), ≥98% | assay 98% | assay - | assay - |
| functional group phosphine | functional group phosphine | functional group - | functional group - |
| Quality Level 300 | Quality Level 300 | Quality Level 100 | Quality Level - |
| storage temp. room temp | storage temp. - | storage temp. - | storage temp. - |
| form solid | form solid | form solid | form - |
Still not finding the right product?
Explore all of our products under BrettPhos
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

