Skip to Content
Merck

Skip To

PMC2012

MorDalPhos

≥98%, ISOM8™

Synonym(s):

MorDalphos, Di(1-adamantyl)-2-morpholinophenylphosphine, Mor-DalPhos

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C30H42NOP
CAS Number:
Molecular Weight:
463.63
UNSPSC Code:
12352005
MDL number:
eCl@ss:
32039090
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ISOM8™

Quality Segment

assay

≥98%

form

solid

purified by

HPLC

reaction suitability

reagent type: ligand, reagent type: ligand
reaction type: Arylations, reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction, reagent type: ligand
reaction type: Coupling Reactions

mp

219-224 °C

application(s)

diagnostic assay manufacturing
life science and biopharma

functional group

phosphine

storage temp.

room temp

SMILES string

C1CN(CCO1)c2ccccc2P([C@@]34C[C@@H]5C[C@@H](C[C@@H](C5)C3)C4)[C@@]67C[C@@H]8C[C@@H](C[C@@H](C8)C6)C7

InChI

1S/C30H42NOP/c1-2-4-28(27(3-1)31-5-7-32-8-6-31)33(29-15-21-9-22(16-29)11-23(10-21)17-29)30-18-24-12-25(19-30)14-26(13-24)20-30/h1-4,21-26H,5-20H2/t21-,22+,23-,24-,25+,26-,29-,30-

InChI key

CCBRRSUORFMQCZ-BVIFAWIJSA-N

General description

Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8 team, we have expertise and documentation to support ISO 9001 industrial and non-GMP pharma production needs, as defined by our M-Clarity™ Program. Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at quality documentation for this product, provided with support of our Elevate Program.

Application

MorDalPhos is an electron-rich, sterically hindered P,N-heterobidentate ligand bridging monophosphines and bisphosphines. It excels in Pd-catalyzed C–N and C–C bond-forming reactions, including Buchwald-Hartwig amination and ammonia monoarylation, with broad functional group tolerance.





Notable Characteristics & Key Uses


  • P,N-Heterobidentate Design: Features a bulky adamantyl-phosphine group paired with a morpholine-containing nitrogen donor, providing a “middle ground” between traditional monophosphines and chelating bisphosphines.
  • High Chemoselectivity: Highly active in chemoselective C–N bond formations, successfully differentiating between closely related amines.
  • Mild Reaction Conditions: Enables reactions to occur under lower catalyst loadings, shorter timeframes, and sometimes under solvent-free or aqueous conditions.
  • Ammonia Monoarylation: Excels in the direct monoarylation of ammonia with aryl chlorides and tosylates — a process historically known to be exceptionally difficult.
  • Heterocycle & Pharmaceutical Synthesis: Frequently applied in the construction of APIs such as MALT1 inhibitors, CNS therapeutics, and anti-inflammatory compounds.
  • Hydrazine & Alkyne Reactions: Used for the monoarylation of hydrazine, acetone derivatives, and in alkyne hydroamination.
  • Functional Group Tolerance: MorDalPhos enables Pd-catalyzed C–N and C–C bond formation with good functional group tolerance, making it highly versatile across diverse synthetic contexts.

Recent Studies and Innovations


  1. Solvent-free amination coupling reactions: MorDalPhos is the enabling ligand for a practical, broad-scope Pd-catalyzed amination platform for triaryl phosphates, with good functional-group tolerance and solvent-free applicability (Zicong Chen et al., 2019).
  2. High Selectivity in Mono-α-Arylation: A highly reactive catalyst system comprising [Pd(2-butenyl)Cl]₂ and MorDalPhos achieves successful mono-α-arylation of aryl and heteroaryl ketones with aryl phosphates. (Xiangmeng Chen et al., 2019).
  3. Ammonia Monoarylation at Room Temperature: MorDalPhos enables selective monoarylation of ammonia, achieving >90% conversion and 76% GC yield of aniline. (Alsabeh et al., 2013).

Legal Information

ISOM8 is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
751618792357
description

≥98%, ISOM8™

description

97%

description

-

reaction suitability

reagent type: ligand
reaction type: Arylations, reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction, reagent type: ligand
reaction type: Coupling Reactions

reaction suitability

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction, reagent type: ligand
reaction type: Arylations

reaction suitability

reaction type: Sonogashira Coupling, reagent type: catalyst
reaction type: Cross Couplings, reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Heck Reaction, reaction type: Hiyama Coupling, reaction type: Negishi Coupling, reaction type: Suzuki-Miyaura Coupling, reaction type: Stille Coupling

functional group

phosphine

functional group

phosphine

functional group

phosphine

assay

≥98%

assay

97%

assay

-

Quality Level

300

Quality Level

100

Quality Level

100

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

form

solid

form

solid

form

solid


Still not finding the right product?

Explore all of our products under MorDalPhos


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library