Skip to Content
Merck

PPMLS

Polyphenol Metabolite Library of Standards

PPMLS, Polyphenol library standards, Polyphenol metabolite library

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

IROA Technologies

suitability

suitable for (metabolite standards), suitable for mass spectrometry

application(s)

advanced drug delivery
lipidomics
metabolomics
microbiology

shipped in

dry ice

storage temp.

−20°C

General description

PPMLS (Polyphenol Metabolite Library of Standards) is a collection of unique high-quality small biochemicals that have a polyphenol structures. Polyphenols are founds in vegetables, cereals, fruits, oils, chocolate, spices, legumes and some beverages like red wine. In many plants, these are secondary metabolites associated with stress defense. Polyphenols are a class of compounds which includes flavonoids, phenolic acids, lignans, and stilbenes. Numerous studies of polyphenolic compounds have shown evidence of their anti-oxidative properties. Consumption of foods containing polyphenolic compounds might limit the risk of degenerative diseases, reduce oxidative stress and inflammation.

Application

Metabolite research. Metabolomic studies. These standards are mostly used to provide retention times and spectra for these key products. Aids in optimizing the mass spectrometry analytical protocols, qualify and quantify mass spectrometry sensitivity. Helps in finding limit of detection for specific analytical parameters.

Features and Benefits

  1. These are high purity (>95%) compounds supplied in an economical ready to use format.
  2. Pre-weighed and supplied in dry form.
  3. Furture each well contains 5 ug per compound enough for multiple injections.
  4. 96- well plate with Alphanumeric assigned positions of products.
  5. A plate map of compounds and descritptors in an excel form.
  6. MLS Discovery - A software tool to support extraction, manipulation and storage of data generated using this library.
To view a complete list of compounds in the library for recent lots, download the plate maps below:

Other Notes

PPMLS contains 80 unique small molecule metabolites covering key metabolic pathways, including the following classes of compounds:
  • Phenols
  • Phenolic terpenes
  • Phenylpropanes
  • Flavonoids, Isoflavonoids. Flavanones and Methoxyflavonols
  • Cinnamic acids and derivatives
  • Quinic acids and derivatives
  • Stilbenes
  • Lignans


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 2 Oral - STOT SE 3

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

120-80-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library