Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
recombinant
expressed in E. coli expressed in
Quality Segment
form
liquid
feature
DNA free, RNase and DNase free (also Nickase free)
packaging
pkg of 0.5 mL
technique(s)
RT-PCR: suitable, cDNA synthesis: suitable
shipped in
dry ice
storage temp.
-10 to -25°C
General description
Ultra pure 10x Reverse Transcriptase Buffer was designed for use in highly-regulated industries in applications that require a high level of performance, lot-to-lot consistency, and high sensitivity without false results from reagent-borne contamination while meeting industry-driven critical regulations. To meet this requirements we have increased the quality level of our products. The M-Clarity Program defines product quality. It provides transparency so that you can choose, with confidence, suitable products that meet your needs with respect to quality and regulatory compliance, transparency, change notification, and documentation.
Application
- Reverse transcription
- First strand cDNA synthesis (up to 7 kb)
- Preparation of cDNA libraries
- RT-PCR and RT-qPCR
- Rapid Amplification of cDNA Ends (RACE)
- RNA-Seq
- For further manufacture
Features and Benefits
- DNA-free
- DNase-free, RNase-free, and nickase-free
- Ideal for high throughput RT-PCR applications
- Compatible with Ultra pure High-temp Reverse Transcriptase
- Enhanced change control and quality agreement
Packaging
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description DNA-free, Nuclease-free 10x RT buffer, designed for further manufacture | description DNA-free, Nuclease-free high-temp RT, designed for further manufacture | description DNA-free, Nuclease-free water | description DNA-free, Nuclease-free 10x PCR reaction buffer with MgCl2, designed for further manufacture |
| shipped in dry ice | shipped in dry ice | shipped in ambient | shipped in dry ice |
| technique(s) RT-PCR: suitable, cDNA synthesis: suitable | technique(s) cDNA synthesis: suitable, RT-PCR: suitable | technique(s) cDNA synthesis: suitable, DNA sequencing: suitable, qPCR: suitable, RT-PCR: suitable | technique(s) cDNA synthesis: suitable, RT-PCR: suitable |
| storage temp. -10 to -25°C | storage temp. −20°C | storage temp. - | storage temp. −20°C |
| form liquid | form liquid | form liquid | form - |
| recombinant expressed in E. coli expressed in | recombinant expressed in E. coli Expressed in, expressed in E. coli expressed in | recombinant - | recombinant - |
| feature DNA free, RNase and DNase free (also Nickase free) | feature RNase and DNase free (also Nickase free), DNA free | feature RNase and DNase free (also Nickase free), DNA free | feature DNA free, RNase and DNase free (also Nickase free) |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

