Skip to Content
Merck

Skip To

A9570

MilliShot® Amphotericin B Ready Made Solution

250 μg/mL in deionized water, 0.2 μm filtered, BioReagent, suitable for cell culture

Synonym(s):

Amphotericin, Fungizone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 x 5 mL
Please contact Customer Service for Availability
767,00 PLN

About This Item

NACRES:
NA.76
eCl@ss:
32040201

767,00 PLN


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

yeast

Quality Segment

sterility

0.2 μm filtered

product line

BioReagent

form

liquid

shelf life

2 yr

storage condition

protect from light

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Amphotericin B is a macrocyclic lactone with a hydrophilic polyhydroxyl chain and a lipophilic polyene based hydrocarbon chain

Application

Amphotericin B solution has been used:
  • as an intraperitoneal injection for the treatment of Aspergillus fumigatus infection in mice
  • as a constituent of aspiration medium for the culture of Chinese Merino ewe ovaries
  • as a constituent of transport medium during the collection of equine corpora lutea
  • as a supplement in RPMI medium for the culture of the mouse-mouse B cell hybridoma 55-6 cell line

Biochem/physiol Actions

Amphotericin B is a polyene antifungal antibiotic from Streptomyces sp. that is clinically used to treat systemic fungal infections. It has a high affinity for sterols, especially ergosterols, of both fungal and bacterial cell membranes. Normal usage in maintenance of antifungal cell cultures is 2.5 mg/L with penicillin and streptomycin used in the medium. Amphotericin B is most effective against fungi and yeast. It forms transmembrane channels by binding to sterols associated with membranes, thereby leading to the leakage of small molecules. Amphotericin B elicits toxicity despite being an effective antibiotic for treating mycoses.

Legal Information

MILLISHOT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G9571A9571A2942
description

250 μg/mL in deionized water, 0.2 μm filtered, BioReagent, suitable for cell culture

description

1 mg/mL in deionized water, 0.2 μm filtered, BioReagent, suitable for cell culture

description

0.2 μm filtered, BioReagent, suitable for cell culture

description

250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

product line

BioReagent

product line

BioReagent

product line

BioReagent

product line

BioReagent

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

200

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

−20°C

biological source

yeast

biological source

microbial

biological source

yeast

biological source

-


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library