Skip to Content
Merck

Skip To

BPS60400

PDE10A2 Assay Kit

Synonym(s):

Assay Kits, PDE10A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41121811
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

technique(s)

activity assay: suitable (Fluorescence Polarization)

UniProt accession no.

shipped in

dry ice

storage temp.

-65 to -96°C

General description

The PDE10A2 Assay Kit is a fluorescence polarization (FP), homogeneous, 96-well assay kit designed for the screening and profiling of PDE10A2 (Phosphodiesterase 10A2) inhibitors. This assay takes advantage of a specific fluorescent phosphate-binding nanoparticle. The kit contains enough purified recombinant PDE10A2, fluorescent probe, PDE assay buffer, Binding Agent, and diluent for 100 reactions. The assay uses a fluorescein-labeled cyclic adenosine monophosphate (cAMP-FAM for PDE10A2), in which the phosphate group is engaged within the cyclic nucleotide. This is a very small molecule that rotates fast (low FP). PDE10A2 catalyzes the hydrolysis of the phosphodiester bond in the cyclic nucleotide and frees the phosphate group. In a second step the free phosphate group is recognized by a specific phosphate-binding nanobead (Binding Agent) leading to the formation of a large complex, with restricted movement (high FP). FP is proportional to PDE10A2 activity.

Application

Study enzyme kinetics and screen small molecule inhibitors for drug discovery in high throughput screening (HTS) applications.
Phosphodiesterases (PDEs) play an important role in the dynamic regulation of the second messengers cAMP (cyclic adenosine monophosphate) and cGMP (cyclic guanosine monophosphate) signaling, by hydrolyzing them. The PDE superfamily is composed of 11 families, with PDE4, 7 and 8 being cAMP-specific hydrolases, and thus regulating positive and negative responses to it. Human PDE10A has multiple biological functions, in a cell-specific context. It is involved in mitochondrial morphology and removal, apoptosis and mouse liver development and hematopoiesis. Additionally, it has been linked to cancer progression, as in colorectal cancer, melanoma, glioma and hepatocellular carcinoma (HCC). Its various functions make it a highly relevant target for the treatment of many disorders, and the use of PDE10A inhibitors will continue to open new therapeutic avenues.
Datasheet and Protocols may be found here.

Features and Benefits

The PDE10A inhibitor screening assay kit comes in a convenient 96-well format, with purified PDE10A enzyme, fluorescently labeled PDE10A substrate (cAMP), binding agent, and PDE assay buffer for 100 enzyme reactions.


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library