Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106514
Biological source:
Mouse x Mouse Hybridoma
Isotype:
IgG1
Technique(s):
cell culture | mammalian: suitable
Myeloma:
NS-1 (P3-X63/Ag8-NS1)
Reactivity:
Anti-L. panamensis-Amastigote & Promastigote
Product Name
LXVIII-4D8-E3, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
Mouse x Mouse Hybridoma
manufacturer/tradename
ECACC
packaging
tube of 5 μg 08041712-DNA-5UG, pkg of vial of cells 08041712-1VL
ImmunoDon
Balb/c mouse spleen
Myeloma
NS-1 (P3-X63/Ag8-NS1)
Reactivity
Anti-L. panamensis-Amastigote & Promastigote
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Immunogen
Membrane from Leishmania (L) mexicana strain MNYC/BZ/62/M379
Application
Detection of Leishmania (L) mexicana complex membranes
Biochem/physiol Actions
Anti-Leishmania (L) mexicana complex
LXVIII-4D8-E3 was developed by immunizing mice with membranes from Leishmania (L) mexicana and fusing the mouse spleen cells with NS-1(P3-X63/Ag8-NS1)
Preparation Note
RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Seed cultures between 1-3x100,000 cells/ml; Saturation density 1,000,000 5% CO2; 37°C
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.