Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
CH3(CH2)11N(O)(CH3)2
CAS Number:
Molecular Weight:
229.40
EC Number:
216-700-6
UNSPSC Code:
12161900
PubChem Substance ID:
Beilstein/REAXYS Number:
1769927
MDL number:
Pricing and availability is not currently available.
description
non-ionic
assay
≥99% (titration)
mol wt
229.40 g/mol
technique(s)
NMR: suitable, enzyme immunoassay: suitable
mp
132-133 °C (lit.)
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCC[N+](C)(C)[O-]
InChI
1S/C14H31NO/c1-4-5-6-7-8-9-10-11-12-13-14-15(2,3)16/h4-14H2,1-3H3
InChI key
SYELZBGXAIXKHU-UHFFFAOYSA-N
General description
N,N-Dimethyldodecylamine N-oxide is a non-denaturing zwitterionic surfactant with a critical micelle concentration (CMC) of approximately 1 mM. It is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.
Other Notes
This product has been replaced by 40234-SIGMA | N,N-Dimethyldodecylamine N-oxide BioXtra, ≥99.0% (NT)
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
PPE (personal protective equipment)
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


