Skip to Content
Merck

Skip To

F4429

FAUC 213

≥98% (HPLC), solid

Synonym(s):

2-[4-(4-Chlorophenyl)piperazin-1-ylmethyl]pyrazolo[1,5-a]pyridine

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid


assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ~24 mg/mL (with warming up to 60 °C), H2O: insoluble

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)N2CCN(CC2)Cc3cc4ccccn4n3

InChI

1S/C18H19ClN4/c19-15-4-6-17(7-5-15)22-11-9-21(10-12-22)14-16-13-18-3-1-2-8-23(18)20-16/h1-8,13H,9-12,14H2

InChI key

DTRXURJDKOYCCD-UHFFFAOYSA-N

Gene Information

Application

FAUC 213 has been used as a selective dopamine receptor D4-R antagonist:
  • to test its effect on the expression of luteinizing hormone in eel pituitary primary cell culture
  • to test its effect on bupropion induced hypophagia and locomotion in rats
  • to test its modulatory effects on rat basolateral amygdala in the presence of dopamine

Biochem/physiol Actions

FAUC 213 displays antipsychotic properties and may be of use in studies related to behavioral neurobiology and neurochemistry.
Highly selective D4 dopamine receptor full antagonist


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters