Skip to Content
Merck

Skip To

PLA0200

Rabbit anti-KPNA3 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

IPOA4, SRP1, SRP1-gamma, SRP1gamma, SRP4, hSRP1, importin alpha 4, importin alpha Q2, importin alpha-3, importin subunit alpha-3, importin subunit alpha-4, importin-alpha-Q2, karyopherin alpha 3, karyopherin alpha 3 (importin alpha 4), karyopherin subunit alpha-3, qip2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
2.180,00 PLN

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-

2.180,00 PLN


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: 1:100- 1:500, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_002258.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... KPNA3(3839)

Immunogen

The epitope recognized by PLA0200 maps to a region between residue 1 and 50 of human karyopherin alpha 3 using the numbering given in entry NP_002258.2 (GeneID 3839).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

KPNA3 (karyopherin alpha 3), also known as importin alpha 4, is a member of the importin alpha family of proteins which includes KPNA1 (importin alpha 5), KPNA2 (importin alpha 1), KPNA3 (importin alpha 4), KPNA4 (importin alpha 3), KPNA5 (importin alpha 6), and KPNA6 (importin alpha 7). The importin alpha family of proteins plays a role in the import of proteins from the cytoplasm to the nucleus. The import of proteins from the cytoplasm to the nucleus involves docking the protein to receptors associated with the nuclear pore complex (NPC) followed by translocation through the nuclear pore. Importin alpha proteins bind the nuclear localization signal (NLS) of cargo proteins and function as adaptor proteins for the nuclear pore receptor KPNB1 (importin beta 1). KPNB1 is responsible for docking the importin-cargo complex to the cytoplasmic side of the NPC for its translocation through the NPC. KPNA3 (Importin alpha 4) and KPNA4 (importin alpha 3) have been reported to mediate the nuclear import of TNF-alpha induced nuclear import of NF-kappaB p50/p65 heterodimers. KPNA3 has also been reported to be responsible for the ligand-independent nuclear translocation of the vitamin D receptor.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA046852SAB5700852SAB3500561
description

Powered by Bethyl Laboratories, Inc.

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

-

description

affinity isolated antibody, buffered aqueous solution

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

human

species reactivity

human

species reactivity

mouse, human

species reactivity

mouse, rat, human

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

UniProt accession no.

O00505

UniProt accession no.

O00505

UniProt accession no.

O00505

UniProt accession no.

O00505


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters