Skip to Content
Merck

Skip To

PLA0215

Rabbit anti-NF-kappaB2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CVID10, DNA-binding factor KBF2, H2TF1, LYT-10, LYT10, NF-kB2, NFKB, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, lymphocyte translocation chromosome 10 protein, nuclear factor Kappa-B, nuclear factor NF-kappa-B p52 subunit, nuclear factor of Kappa light chain gene enhancer in B cells 2, nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100), oncogene Lyt-10, p100, p105, p49/p100, p52, p52/p100 subunit, subunit 2, transcription factor NFKB2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
2.180,00 PLN

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
2

2.180,00 PLN


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_001070962.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... NF-kappaB2(4791)

Immunogen

The epitope recognized by PLA0215 maps to a region between residue 810 and 860 of human nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100) using the numbering given in entry NP_001070962.1 (GeneID 4791).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

The NFKB2 gene encodes a 100 kDa protein that can undergo co-translational processing to also yield a 52 kDa protein. The 52 kDa protein is also known as NF-kappa B2/p52 and is one of five monomeric subunits that are part of the Rel family of proteins that dimerize to form the NF-kappa B (nuclear factor that binds the kappa immunoglobulin light chain gene enhancer) transcription factor. Members of the Rel family of proteins include NF-kappa B1/p50, NF-kappa B2/p52, RelA/p65, c-Rel, and RelB. The 100 kDa product is also known as p100 or I kappa B delta. P100/I kappa B delta has been shown to function as an inhibitor protein that interacts with NF-kappa B and inhibit its DNA binding activity. P100/I kappa B delta is a member of the I kappa B family of proteins which includes I kappa B-beta, I kappa B-delta/NF-kappa B2/p100, I kappa B-gamma/NF-kappa B1/p105, I kappa B-epsilon, I kappa B-zeta, and Bcl3. The classical description of the NF-kappa B transcription factor is a transcription factor that reprograms gene expression of the immune system in response to injury, infection, and stress.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5702194ABS1515SAB4301419
description

Powered by Bethyl Laboratories, Inc.

description

-

description

from rabbit, purified by affinity chromatography

description

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

-

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

polyclonal

clone

4K3V3, monoclonal

clone

polyclonal

clone

polyclonal

species reactivity

mouse, human

species reactivity

rat, human

species reactivity

human

species reactivity

rat, human, mouse

antibody form

affinity purified immunoglobulin

antibody form

-

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library