Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Please contact Customer Service for Availability | 2.470,00 PLN |
About This Item
sample type urine
sample type serum
sample type plasma
2.470,00 PLN
Quality Segment
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable, capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 600 pg/mL
standard curve range: 0.68-500 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... TIE1(7075)
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
Please type the word sample in the text box provided for lot number.
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma, cell culture supernatant and urine | description - | description for serum, plasma, cell culture supernatants and urine | description for serum, plasma, cell culture supernatant and urine |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| Gene Information human ... TIE1(7075) | Gene Information human ... TIE1(7075) | Gene Information human ... TEK(7010) | Gene Information human ... ANGPT1(284) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <10% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type cell culture supernatant(s) | input - | input sample type urine | input sample type urine |
| Quality Level 100 | Quality Level - | Quality Level 100 | Quality Level - |
| detection method colorimetric | detection method - | detection method - | detection method colorimetric |
Signal Word
Danger
Storage Class
10 - Combustible liquids
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




