Skip to Content
Merck

Skip To

RAB0969

Phosphotyrosine ITK / Tyrosine-protein Kinase ITK/TSK ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Please contact Customer Service for Availability
2.490,00 PLN

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... ITK(3702)
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

2.490,00 PLN


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ITK(3702)

General description

The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-ITK.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0966RAB0972RAB0963
description

-

description

-

description

-

description

-

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

Gene Information

human ... ITK(3702)

Gene Information

human ... HCK(3055)

Gene Information

human ... JAK3(3718)

Gene Information

human ... FGR(2268)

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters