Skip to Content
Merck

Skip To

SCP0010

Adrenomedullin 11-50 rat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 μg
Please contact Customer Service for Availability
2.020,00 PLN
1 mg
Please contact Customer Service for Availability
3.370,00 PLN

About This Item

Empirical Formula (Hill Notation):
C194H304N58O59S4
Molecular Weight:
4521.10
NACRES:
NA.32
UNSPSC Code:
12352200

2.020,00 PLN


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Application

Adrenomedullin 11-50 rat is a peptide fragment of adrenomedullin that possesses systemic vasodilator activity at the level of calcitonin gene-related peptide-1 (CGRP1) receptor activation. Adrenomedullin and its peptide may be used in bone growth studies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3830A2327A3832
description

-

description

≥95% (HPLC)

description

≥90% (HPLC), powder

description

≥97% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥90% (HPLC)

assay

≥97% (HPLC)

form

lyophilized

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage condition

protect from light

storage condition

-

storage condition

-

storage condition

-

composition

Peptide Content, ≥70%

composition

-

composition

-

composition

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters