Skip to Content
Merck

Skip To

SML2291

S119-8

≥98% (HPLC)

Synonym(s):

4-(tert-butyl)-N-(4-(phenylamino)phenyl)benzamide, Benzamide, 4-(1,1-dimethylethyl)-N-[4-(phenylamino)phenyl]

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
1.300,00 PLN

About This Item

Empirical Formula (Hill Notation):
C23H24N2O
CAS Number:
Molecular Weight:
344.45
UNSPSC Code:
12352200
NACRES:
NA.77
eCl@ss:
32160703
Assay:
≥98% (HPLC)
Form:
powder

1.300,00 PLN


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(C(C)(C)C)C=C1)NC2=CC=C(NC3=CC=CC=C3)C=C2

InChI key

NEPKMQDGCTXOPG-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of viral nucleoprotein of influenza A and B
S119-8 is an inhibitor of viral nucleoprotein of influenza A and B. It showed synergistic effects with oseltamivir.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1271MAB8259MAB8258B5
description

≥98% (HPLC)

description

≥98% (HPLC)

description

clone B2, Chemicon®, from mouse

description

clone A3, Chemicon®, from mouse

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

-

assay

-

form

powder

form

powder

form

-

form

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

-

solubility

-

color

white to beige

color

white to beige

color

-

color

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters