Skip to Content
Merck

Skip To

SML4100

Vidofludimus

≥98% (HPLC)

Synonym(s):

IMU-838, 2-((3-Fluoro-3′-methoxy-[1,1′-biphenyl]-4-yl)carbamoyl)cyclopent-1-enecarboxylic acid, 2-[[(3-Fluoro-3′-methoxy[1,1′-biphenyl]-4-yl)amino]carbonyl]-1-cyclopentene-1-carboxylic acid, 2-[[2-Fluoro-4-(3-methoxyphenyl)phenyl]carbamoyl]cyclopentene-1-carboxylic acid, 4SC-101, IMU 838, IMU838, SC12267

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Please contact Customer Service for Availability
473,00 PLN
50 mg
Please contact Customer Service for Availability
1.520,00 PLN

About This Item

Empirical Formula (Hill Notation):
C20H18FNO4
CAS Number:
Molecular Weight:
355.36
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder

473,00 PLN


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to yellow

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

Fc1c(ccc(c1)c3cc(ccc3)OC)NC(=O)C2=C(CCC2)C(=O)O

InChI

1S/C20H18FNO4/c1-26-14-5-2-4-12(10-14)13-8-9-18(17(21)11-13)22-19(23)15-6-3-7-16(15)20(24)25/h2,4-5,8-11H,3,6-7H2,1H3,(H,22,23)(H,24,25)

InChI key

XPRDUGXOWVXZLL-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, potent dihydroorotate dehydrogenase (DHODH) inhibitor in cultures and in vivo.
Vidofludimus (4SC-101) is an orally active, potent dihydroorotate dehydrogenase (DHODH) inhibitor (human DHODH IC50 = 134 nM; IC50 = 13 µM against PHA-stimulated lymphocytes proliferation) that inhibits PHA-induced IL-17 secretion from human PBMCs (IC50 = 6 µM) and exhibits therapeutic efficacy in murine models of colitis (50-100 mg/kg/d via intragastric administration; chronic dextran sodium sulfate-induced and acute TNBS-induced colitis) and a rat model of renal transplantation in vivo (20 mg/kg/d p.o.).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABC138HPA010123HPA011942
description

≥98% (HPLC)

description

from rabbit, purified by affinity chromatography

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

form

powder

form

-

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

-10 to -25°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

solubility

-

color

white to yellow

color

-

color

-

color

-


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library