Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C38H36F6N6O4·C4H6O4
CAS Number:
Molecular Weight:
872.81
UNSPSC Code:
12352200
MDL number:
eCl@ss:
32160701
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Quality Segment
assay
≥98% (HPLC)
form
powder
drug control
USDEA Schedule V
storage condition
desiccated
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Lasmiditan (COL-144 and LY573144) is an orally active, potent and selective high-affinity serotonin receptor 5-HT1F agonist (5-HT1F/1E Ki = 2.21/594 nM; 5-HT1A/B/C Ki >1 μM; 5-HT2A/B/C Ki >2 μM; 5-HT6/7 Ki >3 μM). Lasmiditan induces mitochondrial biogenesis (MB) in human glomerular endothelial cells and mouse glomerular endothelial cells (100 nM for 72h). Unlike the triptans, Lasmiditan is void of vasoconstrictor activity toward isolated human coronary arteries (up to 100 μM) or in anaesthetized female beagle dogs in vivo (4.13-11.13 mg/kg i.v.)
Disclaimer
Hygroscopic, Store with desiccant
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


