Skip to Content
Merck

Skip To

1A00580

USP

4-Fluoro Risperidone (Cold Shipment Required)

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

(3-(2-(4-(4-fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl)ethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C23H27FN4O2
Molecular Weight:
410.48
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

API family

risperidone

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.
4-Fluoro Risperidone (Cold Shipment Required) is a USP Pharmaceutical Analytical Impurity (PAI).

USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.

Associated Drug Substance: Risperidone

Therapeutic Area: Psychiatrics.

For more information about this PAI, visit here.

Application

4-Fluoro Risperidone (Cold Shipment Required) (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1A005201A005901A11840
description

Pharmaceutical Analytical Impurity (PAI)

description

Pharmaceutical Analytical Impurity (PAI)

description

Pharmaceutical Analytical Impurity (PAI)

description

pharmaceutical analytical impurity (PAI)

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical small molecule

API family

risperidone

API family

risperidone

API family

risperidone

API family

-

agency

USP

agency

USP

agency

USP

agency

USP


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters