Skip to Content
Merck

Skip To

1A02430

USP

Chlorobutoxyquinolinone

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

Chlorobutoxyquinolinone, (7-(4-chlorobutoxy)-3,4-dihydroquinolin-2(1H)-one)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
5.350,00 PLN

About This Item

Empirical Formula (Hill Notation):
C13H16ClNO2
CAS Number:
Molecular Weight:
253.72
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107

5.350,00 PLN


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Chlorobutoxyquinolinone is a USP Pharmaceutical Analytical Impurity (PAI).


USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.


Associated Drug Substance: Aripiprazole


Therapeutic Area: Psychiatrics

Application

Chlorobutoxyquinolinone is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR92781A025201A02470
description

Pharmaceutical Analytical Impurity (PAI)

description

certified reference material, TraceCERT®

description

Pharmaceutical Analytical Impurity (PAI)

description

Pharmaceutical Analytical Impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

grade

TraceCERT®, certified reference material

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical

format

neat

format

-

format

neat

format

neat

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

2-8°C

agency

USP

agency

(Not traceable to any primary pharmacopeia standard)

agency

USP

agency

USP


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library