Skip to Content
Merck

Skip To

1A03400

USP

Bortezomib L-Phenylalanyl Analog

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

Bortezomib L-Phenylalanyl Analog, (((R)-3-METHYL-1-((S)-3-PHENYL-2-((S)-3-PHENYL-2-(PYRAZINE-2-CARBOXAMIDO)PROPANAMIDO)PROPANAMIDO)BUTYL)BORONIC ACID)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C28H34BN5O5
CAS Number:
Molecular Weight:
531.41
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

API family

bortezomib

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Bortezomib L-Phenylalanyl Analog is a USP Pharmaceutical Analytical Impurity (PAI).


USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.


Associated Drug Substance: Bortezomib


Therapeutic Area: Oncology

Application

Bortezomib L-Phenylalanyl Analog (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
FLUH9C59ED48FLUH9C33EC37
description

Pharmaceutical Analytical Impurity (PAI)

description

-

description

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical analytical impurity (PAI)

grade

-

grade

-

application(s)

pharmaceutical
pharmaceutical
pharmaceutical small molecule

application(s)

-

application(s)

-

format

neat

format

-

format

-

agency

USP

agency

-

agency

-

API family

bortezomib

API family

-

API family

-


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Gastro-intestinal system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library