Merck
  • Phagocytosis-dependent activation of a TLR9-BTK-calcineurin-NFAT pathway co-ordinates innate immunity to Aspergillus fumigatus.
Phagocytosis-dependent activation of a TLR9-BTK-calcineurin-NFAT pathway co-ordinates innate immunity to Aspergillus fumigatus.
EMBO molecular medicine (2015-02-01)
Susanne Herbst, Anand Shah, Maria Mazon Moya, Vanessa Marzola, Barbara Jensen, Anna Reed, Mark A Birrell, Shinobu Saijo, Serge Mostowy, Sunil Shaunak, Darius Armstrong-James
ABSTRACT
Transplant recipients on calcineurin inhibitors are at high risk of invasive fungal infection. Understanding how calcineurin inhibitors impair fungal immunity is a key priority for defining risk of infection. Here, we show that the calcineurin inhibitor tacrolimus impairs clearance of the major mould pathogen Aspergillus fumigatus from the airway, by inhibiting macrophage inflammatory responses. This leads to defective early neutrophil recruitment and fungal clearance. We confirm these findings in zebrafish, showing an evolutionarily conserved role for calcineurin signalling in neutrophil recruitment during inflammation. We find that calcineurin-NFAT activation is phagocytosis dependent and collaborates with NF-κB for TNF-α production. For yeast zymosan particles, activation of macrophage calcineurin-NFAT occurs via the phagocytic Dectin-1-spleen tyrosine kinase pathway, but for A. fumigatus, activation occurs via a phagosomal TLR9-dependent and Bruton's tyrosine kinase-dependent signalling pathway that is independent of MyD88. We confirm the collaboration between NFAT and NF-κB for TNF-α production in primary alveolar macrophages. These observations identify inhibition of a newly discovered macrophage TLR9-BTK-calcineurin-NFAT signalling pathway as a key immune defect that leads to organ transplant-related invasive aspergillosis.
Product Description
Supelco
Water, for TOC analysis
Pricing and availability is not currently available.
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Supelco
D-(+)-Glucose, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Pricing and availability is not currently available.
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Water, tested according to Ph. Eur.
Pricing and availability is not currently available.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Piceatannol, powder
Pricing and availability is not currently available.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Pricing and availability is not currently available.
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Water, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Pricing and availability is not currently available.